Product Details

SNP ID
rs146655157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58568658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGGACAGGGCCAGCCAATCA[A/G]CTGCAAATAAAATTTGGTTGGTAAC
Phenotype
MIM: 605792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX14 PubMed Links

Gene Details

Gene
TEX14
Gene Name
testis expressed 14, intercellular bridge forming factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201457.1 Intron NP_001188386.1
NM_031272.4 Intron NP_112562.3
NM_198393.3 Intron NP_938207.2
XM_011525029.2 Intron XP_011523331.1
XM_017024860.1 Intron XP_016880349.1
XM_017024861.1 Intron XP_016880350.1

View Full Product Details