Product Details
- SNP ID
-
rs146805764
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44848189 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACGCTGGTGGGTACCACCTGAC[A/G]ACGAAGACTGTGTGTCTGAGAAGCT
- Phenotype
-
MIM: 603892
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFTUD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12164] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFTUD2
- Gene Name
- elongation factor Tu GTP binding domain containing 2
There are no transcripts associated with this gene.
- Gene
- HIGD1B
- Gene Name
- HIG1 hypoxia inducible domain family member 1B
View Full Product Details