Product Details

SNP ID
rs146845700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55267914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGACCTGGAGGAGTTTTTGTC[A/G]GAAAATGGCATTCCCCCCAGCCCAT
Phenotype
MIM: 142385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLF PubMed Links

Gene Details

Gene
HLF
Gene Name
HLF, PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002126.4 258 Silent Mutation TCA,TCG S93S NP_002117.1
XM_005257269.2 258 Silent Mutation TCA,TCG S93S XP_005257326.1
XM_011524705.1 258 Silent Mutation TCA,TCG S8S XP_011523007.1
XM_017024556.1 258 Silent Mutation TCA,TCG S8S XP_016880045.1

View Full Product Details