Product Details

SNP ID
rs146866423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56834830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGCGGCCGGCGCCGGGCTCGC[C/T]CTCCGAGGGCCTGTTTGCGGACGGG
Phenotype
MIM: 601440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085430.2 215 Intron NP_001078899.2
XM_011524735.2 215 Intron XP_011523037.1
Gene
DGKE
Gene Name
diacylglycerol kinase epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003647.2 215 Missense Mutation CCC,CTC P12L NP_003638.1
XM_011525394.2 215 Missense Mutation CCC,CTC P30L XP_011523696.1
XM_011525395.2 215 Missense Mutation CCC,CTC P30L XP_011523697.1
XM_011525396.2 215 Missense Mutation CCC,CTC P30L XP_011523698.1
XM_017025243.1 215 Missense Mutation CCC,CTC P12L XP_016880732.1
XM_017025244.1 215 Missense Mutation CCC,CTC P30L XP_016880733.1
XM_017025245.1 215 Missense Mutation CCC,CTC P30L XP_016880734.1

View Full Product Details