Product Details

SNP ID
rs146993812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48577006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGAGCGCTTGGGCTCCCCGCCGG[A/C]GTAATTGGGGTTTACTGGACACACA
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 534 Intron NP_002137.4
XM_005257277.3 534 Intron XP_005257334.1
XM_005257280.3 534 Intron XP_005257337.1
XM_005257282.3 534 Intron XP_005257339.1
XM_006721854.2 534 Intron XP_006721917.1
XM_011524707.2 534 Intron XP_011523009.1
XM_011524708.2 534 Intron XP_011523010.1
XM_011524710.1 534 Intron XP_011523012.1
XM_011524719.1 534 Intron XP_011523021.1
XM_011524720.2 534 Intron XP_011523022.1
XM_011524721.2 534 Intron XP_011523023.1
XM_011524726.2 534 Intron XP_011523028.1
XM_017024557.1 534 Intron XP_016880046.1
XM_017024558.1 534 Intron XP_016880047.1
XM_017024559.1 534 Intron XP_016880048.1
XM_017024560.1 534 Intron XP_016880049.1
XM_017024561.1 534 Intron XP_016880050.1
XM_017024562.1 534 Intron XP_016880051.1
XM_017024563.1 534 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 534 Missense Mutation GCC,TCC A158S NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details