Product Details

SNP ID
rs147169833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42126774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACCTATTGCCATGAAGATTTC[A/G]TTCACGTTCATTGCAGTCTTTGCTG
Phenotype
MIM: 608344 MIM: 602301 MIM: 604037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPB9 PubMed Links

Gene Details

Gene
HSPB9
Gene Name
heat shock protein family B (small) member 9
There are no transcripts associated with this gene.

Gene
KAT2A
Gene Name
lysine acetyltransferase 2A
There are no transcripts associated with this gene.

Gene
RAB5C
Gene Name
RAB5C, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252039.1 695 Silent Mutation AAC,AAT N205N NP_001238968.1
NM_004583.3 695 Silent Mutation AAC,AAT N172N NP_004574.2
NM_201434.2 695 Silent Mutation AAC,AAT N172N NP_958842.1
XM_011525090.1 695 Silent Mutation AAC,AAT N172N XP_011523392.1
XM_011525091.1 695 Silent Mutation AAC,AAT N172N XP_011523393.1

View Full Product Details