Product Details

SNP ID
rs147289705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78175083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCGAGCCTCTTGGTATAGGC[G/T]GCTTCCCGGAAGCACTCCATGCACA
Phenotype
MIM: 603926 MIM: 188300
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C17orf99 PubMed Links

Gene Details

Gene
C17orf99
Gene Name
chromosome 17 open reading frame 99
There are no transcripts associated with this gene.

Gene
SYNGR2
Gene Name
synaptogyrin 2
There are no transcripts associated with this gene.

Gene
TK1
Gene Name
thymidine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003258.4 694 Silent Mutation NP_003249.3
XM_017024992.1 694 Silent Mutation XP_016880481.1

View Full Product Details