Product Details

SNP ID
rs147766716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8254257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGATGTTGCTCGGGAGTCCTG[A/G]CTCCTTCCTGGCTCCAATGACCTGC
Phenotype
MIM: 613129 MIM: 602133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTC1 PubMed Links

Gene Details

Gene
CTC1
Gene Name
CTS telomere maintenance complex component 1
There are no transcripts associated with this gene.

Gene
PFAS
Gene Name
phosphoribosylformylglycinamidine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012393.2 375 Nonsense Mutation TGA,TGG *78W NP_036525.1
XM_006721546.2 375 Nonsense Mutation TGA,TGG *78W XP_006721609.1
XM_017024760.1 375 UTR 5 XP_016880249.1

View Full Product Details