Product Details

SNP ID
rs147883276
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41034389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACTGTCCACAGTAGGATGGGC[A/G]GCAGCAGGAGGCCTCGGCGTGGTGC
Phenotype
MIM: 608819 MIM: 608820 MIM: 608821 MIM: 608822
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRTAP1-1 PubMed Links
Additional Information
For this assay, SNP(s) [rs12936959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRTAP1-1
Gene Name
keratin associated protein 1-1
There are no transcripts associated with this gene.

Gene
KRTAP1-3
Gene Name
keratin associated protein 1-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030966.1 467 Missense Mutation CGC,TGC R145C NP_112228.1
Gene
KRTAP1-4
Gene Name
keratin associated protein 1-4
There are no transcripts associated with this gene.

Gene
KRTAP1-5
Gene Name
keratin associated protein 1-5
There are no transcripts associated with this gene.

View Full Product Details