Product Details
- SNP ID
-
rs148401792
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:77883240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGTGCTGGCCCCTGCAGCAGAG[G/T]CAGAGGCCCAGGCAGGCGGGGACCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ45079
PubMed Links
Gene Details
- Gene
- FLJ45079
- Gene Name
- FLJ45079 protein
There are no transcripts associated with this gene.
View Full Product Details