Product Details

SNP ID
rs148892789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47953109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGCTGTTGGGGCCTCCAGCGTC[A/T]GGCCTCGGGGGTTCTGCATCTCCCT
Phenotype
MIM: 603287
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
There are no transcripts associated with this gene.

Gene
PRR15L
Gene Name
proline rich 15 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024320.3 406 Silent Mutation CCA,CCT P42P NP_077296.1
XM_005257663.4 406 Silent Mutation CCA,CCT P42P XP_005257720.1
XM_005257664.2 406 Silent Mutation CCA,CCT P42P XP_005257721.1
XM_005257665.1 406 Silent Mutation CCA,CCT P42P XP_005257722.1
XM_005257666.4 406 Silent Mutation CCA,CCT P42P XP_005257723.1

View Full Product Details