Product Details

SNP ID
rs148893862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7654148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAGTCCTCAACTACCCCAAAC[A/G]TGCCTGCCAATTCAACCGGACCCAG
Phenotype
MIM: 182331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1B2 PubMed Links

Gene Details

Gene
ATP1B2
Gene Name
ATPase Na+/K+ transporting subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303263.1 1072 Missense Mutation CAT,CGT H66R NP_001290192.1
NM_001678.4 1072 Missense Mutation CAT,CGT H148R NP_001669.3

View Full Product Details