Product Details
- SNP ID
-
rs148902716
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:46769942 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCGCCAGGCGGCCCCTTATGATG[C/T]GAGTCACAGCCGCAAATGGTGGAGG
- Phenotype
-
MIM: 165330
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT3
PubMed Links
Gene Details
- Gene
- WNT3
- Gene Name
- Wnt family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030753.4 |
592 |
Silent Mutation |
TCA,TCG |
S143S |
NP_110380.1 |
View Full Product Details