Product Details

SNP ID
rs149051361
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4631647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACGCTGGGCCGCAGGTACTCGT[A/G]GGGCATGTCCAGCTTTGCATTCCGG
Phenotype
MIM: 152392
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ALOX15 PubMed Links

Gene Details

Gene
ALOX15
Gene Name
arachidonate 15-lipoxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001140.3 1956 Missense Mutation NP_001131.3

View Full Product Details