Product Details
- SNP ID
-
rs149312249
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4797745 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCCTCTTTAAGGAGATGTGTTA[C/G]CGATACCGGGAAGACCTGATGGCGG
- Phenotype
-
MIM: 602384
MIM: 600307
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GLTPD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2304975] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLTPD2
- Gene Name
- glycolipid transfer protein domain containing 2
There are no transcripts associated with this gene.
- Gene
- LOC107984973
- Gene Name
- atherin-like
There are no transcripts associated with this gene.
- Gene
- PLD2
- Gene Name
- phospholipase D2
There are no transcripts associated with this gene.
- Gene
- PSMB6
- Gene Name
- proteasome subunit beta 6
View Full Product Details