Product Details
- SNP ID
-
rs149371094
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8312217 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGCACCAACCCCAATGTGCACC[C/G]CCATCTTCTGGGAGCCCCCAGCTGC
- Phenotype
-
MIM: 608504
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3744651] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF15
- Gene Name
- Rho guanine nucleotide exchange factor 15
View Full Product Details