Product Details

SNP ID
rs149396223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5105908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCATAAGCTTTCCCACATTCTTT[A/G]CATATAAAAGGTTTCTCTCCACTGT
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1369 Silent Mutation TGC,TGT C408C NP_001307881.1
NM_001320953.1 1369 Silent Mutation TGC,TGT C390C NP_001307882.1
NM_001320954.1 1369 Silent Mutation TGC,TGT C390C NP_001307883.1
NM_001320955.1 1369 Silent Mutation TGC,TGT C389C NP_001307884.1
NM_014519.3 1369 Silent Mutation TGC,TGT C417C NP_055334.2
XM_011524006.2 1369 Silent Mutation TGC,TGT C369C XP_011522308.1
XM_017025021.1 1369 Silent Mutation TGC,TGT C417C XP_016880510.1
XM_017025022.1 1369 Silent Mutation TGC,TGT C341C XP_016880511.1
XM_017025023.1 1369 Silent Mutation TGC,TGT C332C XP_016880512.1

View Full Product Details