Product Details

SNP ID
rs149635149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75847545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGGTTCCATGGGCCCAGGGTA[A/G]GGCGGTGGTGGGGGCTGCACGTATC
Phenotype
MIM: 608897 MIM: 606962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UNC13D PubMed Links

Gene Details

Gene
UNC13D
Gene Name
unc-13 homolog D
There are no transcripts associated with this gene.

Gene
WBP2
Gene Name
WW domain binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012478.3 506 Silent Mutation CCC,CCT P199P NP_036610.2
XM_011524580.2 506 Silent Mutation CCC,CCT P154P XP_011522882.1

View Full Product Details