Product Details

SNP ID
rs150281743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62967750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGGCATTTTGAGGGGTTGGAG[A/C]CAGTATCTATCACAATGGTTAATTA
Phenotype
MIM: 615047
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TANC2 PubMed Links

Gene Details

Gene
TANC2
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025185.3 Intron NP_079461.2
XM_005257203.4 Intron XP_005257260.1
XM_006721806.3 Intron XP_006721869.1
XM_006721807.3 Intron XP_006721870.1
XM_006721810.3 Intron XP_006721873.1
XM_006721811.3 Intron XP_006721874.1
XM_011524597.2 Intron XP_011522899.1
XM_011524598.2 Intron XP_011522900.1
XM_011524599.1 Intron XP_011522901.1
XM_011524600.2 Intron XP_011522902.1
XM_011524601.2 Intron XP_011522903.1
XM_011524602.2 Intron XP_011522904.1
XM_017024429.1 Intron XP_016879918.1
XM_017024430.1 Intron XP_016879919.1
XM_017024431.1 Intron XP_016879920.1
XM_017024432.1 Intron XP_016879921.1

View Full Product Details