Product Details

SNP ID
rs150669707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58693895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGATTTTCATTTCCTAAATGCT[A/G]TCATCACCATTGTCTTTTATTAGCC
Phenotype
MIM: 602774 MIM: 605792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51C PubMed Links

Gene Details

Gene
RAD51C
Gene Name
RAD51 paralog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002876.3 Intron NP_002867.1
NM_058216.2 Intron NP_478123.1
XM_006722001.3 Intron XP_006722064.1
XM_006722002.3 Intron XP_006722065.1
XM_006722004.3 Intron XP_006722067.1
XM_006722005.3 Intron XP_006722068.1
XM_011525092.2 Intron XP_011523394.1
XM_011525093.2 Intron XP_011523395.1
XM_011525094.2 Intron XP_011523396.1
XM_017024914.1 Intron XP_016880403.1
XM_017024915.1 Intron XP_016880404.1
XM_017024916.1 Intron XP_016880405.1
XM_017024917.1 Intron XP_016880406.1
XM_017024918.1 Intron XP_016880407.1
XM_017024919.1 Intron XP_016880408.1
Gene
TEX14
Gene Name
testis expressed 14, intercellular bridge forming factor
There are no transcripts associated with this gene.

View Full Product Details