Product Details

SNP ID
rs150692942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19739009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGACTCACCCACGCCCCCGAA[A/G]GGCAGAGAGTGCAAGGTGATGTGGA
Phenotype
MIM: 100660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3A1 PubMed Links

Gene Details

Gene
ALDH3A1
Gene Name
aldehyde dehydrogenase 3 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000691.4 1147 Silent Mutation CCC,CCT P401P NP_000682.3
NM_001135167.1 1147 Silent Mutation CCC,CCT P401P NP_001128639.1
NM_001135168.1 1147 Silent Mutation CCC,CCT P401P NP_001128640.1
XM_005256522.1 1147 Silent Mutation CCC,CCT P518P XP_005256579.1
XM_005256523.2 1147 Silent Mutation CCC,CCT P518P XP_005256580.1
XM_005256524.3 1147 Silent Mutation CCC,CCT P518P XP_005256581.1
XM_011523730.1 1147 Silent Mutation CCC,CCT P328P XP_011522032.1
XM_011523731.1 1147 Silent Mutation CCC,CCT P328P XP_011522033.1

View Full Product Details