Product Details

SNP ID
rs150762833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76144810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGGAGCGATCCTCCAGCCCTA[C/T]CCCAAACTGCAGGTTTCCAATTGGT
Phenotype
MIM: 602291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXJ1 PubMed Links

Gene Details

Gene
FOXJ1
Gene Name
forkhead box J1
There are no transcripts associated with this gene.

Gene
RNF157
Gene Name
ring finger protein 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052916.2 2534 UTR 3 NP_443148.1
XM_005257007.4 2534 UTR 3 XP_005257064.1
XM_011524273.2 2534 Intron XP_011522575.1
XM_017024117.1 2534 Intron XP_016879606.1
XM_017024118.1 2534 Intron XP_016879607.1
XM_017024119.1 2534 UTR 3 XP_016879608.1
XM_017024120.1 2534 Intron XP_016879609.1
Gene
RNF157-AS1
Gene Name
RNF157 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details