Product Details

SNP ID
rs150843841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1707753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCCCCACCTCCCCCAGCGAG[A/G]GGCCCATGGCTTCTCTCTAGTCTTT
Phenotype
MIM: 612077 MIM: 614218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR22 PubMed Links

Gene Details

Gene
MIR22
Gene Name
microRNA 22
There are no transcripts associated with this gene.

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
TLCD2
Gene Name
TLC domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164407.1 938 UTR 3 NP_001157879.1
Gene
WDR81
Gene Name
WD repeat domain 81
There are no transcripts associated with this gene.

View Full Product Details