Product Details

SNP ID
rs151042252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7309645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGAAGCCTCTTAAAATGGCAGAT[A/G]ACTTGGACTTCGAGACAGGAGATGC
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143760.1 226 Intron NP_001137232.1
NM_001143761.1 226 Intron NP_001137233.1
NM_001143762.1 226 Intron NP_001137234.1
NM_001970.4 226 Intron NP_001961.1
XM_005256509.2 226 Intron XP_005256566.1
XM_011523710.2 226 Missense Mutation ATA,ATG I1M XP_011522012.1
XM_011523711.2 226 Missense Mutation ATA,ATG I1M XP_011522013.1
XM_011523712.2 226 Missense Mutation ATA,ATG I1M XP_011522014.1
XM_011523713.2 226 Missense Mutation ATA,ATG I1M XP_011522015.1
XM_017024300.1 226 Missense Mutation ATA,ATG I1M XP_016879789.1
XM_017024301.1 226 Missense Mutation ATA,ATG I1M XP_016879790.1
Gene
GPS2
Gene Name
G protein pathway suppressor 2
There are no transcripts associated with this gene.

Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details