Product Details
- SNP ID
-
rs138690991
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:60371580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGTGGAGGAAGAATGGGGCCCAG[C/G]AGACAACAAAGACGCCAATCAGGAT
- Phenotype
-
MIM: 155541
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MC4R
PubMed Links
Gene Details
- Gene
- MC4R
- Gene Name
- melanocortin 4 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005912.2 |
1189 |
Missense Mutation |
TCC,TGC |
S257C |
NP_005903.2 |
View Full Product Details