Product Details

SNP ID
rs138791943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63331833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTCTCTCTGCATCATGCAGCG[A/T]CGAACTATGCTGTCAAAACTTCCAA
Phenotype
MIM: 136440
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KDSR PubMed Links

Gene Details

Gene
KDSR
Gene Name
3-ketodihydrosphingosine reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002035.2 968 Silent Mutation CGA,CGT R316R NP_002026.1
XM_005266677.2 968 Silent Mutation CGA,CGT R282R XP_005266734.1
XM_017025690.1 968 Silent Mutation CGA,CGT R239R XP_016881179.1

View Full Product Details