Product Details

SNP ID
rs140858508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35336768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTCAAATGCATATTCAAATTA[A/G]GTAAAAGGAAATAAGGGTAAATTTT
Phenotype
MIM: 194534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 1749 Intron NP_001295052.1
NM_006965.3 1749 UTR 3 NP_008896.2
XM_005258341.3 1749 UTR 3 XP_005258398.1

View Full Product Details