Product Details

SNP ID
rs141295699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54903029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAATAAGCAAGCAATAAAAATG[C/T]TTCCAAAAATACCATGAAGCGTAAA
Phenotype
MIM: 616909 MIM: 603869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC68 PubMed Links

Gene Details

Gene
CCDC68
Gene Name
coiled-coil domain containing 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143829.1 2613 UTR 3 NP_001137301.1
NM_025214.2 2613 UTR 3 NP_079490.1
XM_006722552.3 2613 UTR 3 XP_006722615.1
XM_011526198.2 2613 UTR 3 XP_011524500.1
XM_011526199.2 2613 UTR 3 XP_011524501.1
XM_011526200.2 2613 UTR 3 XP_011524502.1
XM_011526201.2 2613 UTR 3 XP_011524503.1
XM_011526202.1 2613 UTR 3 XP_011524504.1
XM_011526203.2 2613 UTR 3 XP_011524505.1
XM_011526204.2 2613 Intron XP_011524506.1
XM_017026011.1 2613 UTR 3 XP_016881500.1
Gene
RAB27B
Gene Name
RAB27B, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details