Product Details

SNP ID
rs142020276
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74253623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATAGGCGATACATCAAGGCGAC[A/G]GCCACTGCAGAGATGGCAGGGATCA
Phenotype
MIM: 613218
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CYB5A PubMed Links

Gene Details

Gene
CYB5A
Gene Name
cytochrome b5 type A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190807.2 477 Silent Mutation GCC,GCT A112A NP_001177736.1
NM_001914.3 477 UTR 3 NP_001905.1
NM_148923.3 477 Silent Mutation GCC,GCT A122A NP_683725.1
XM_011525835.2 477 Intron XP_011524137.1

View Full Product Details