Product Details

SNP ID
rs142410803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31068905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGAAAACTCACTTCACCAAGAC[A/G]GGGCTGAGTAAAACTGTGCCACTCC
Phenotype
MIM: 125645
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSC2 PubMed Links

Gene Details

Gene
DSC2
Gene Name
desmocollin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004949.4 2958 Missense Mutation NP_004940.1
NM_024422.4 2958 Missense Mutation NP_077740.1
XM_005258206.4 2958 Missense Mutation XP_005258263.1

View Full Product Details