Product Details
- SNP ID
-
rs142520163
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:34507435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTATTATTATTAATTTCTTTTGAT[A/G]TAAAAATTATGCATTAAAAATAAAT
- Phenotype
-
MIM: 601239
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DTNA
PubMed Links
Gene Details
- Gene
- DTNA
- Gene Name
- dystrobrevin alpha
View Full Product Details