Product Details

SNP ID
rs144179896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31624701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAACTGGAAATAATAAAAGTG[C/T]TGTTCACCGTGAATTTAAAAAAAAT
Phenotype
MIM: 604017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALT6 PubMed Links

Gene Details

Gene
B4GALT6
Gene Name
beta-1,4-galactosyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004775.3 2405 UTR 3 NP_004766.2
XM_005258387.4 2405 UTR 3 XP_005258444.1
XM_006722578.3 2405 UTR 3 XP_006722641.1
XM_006722579.3 2405 UTR 3 XP_006722642.1
XM_017026088.1 2405 UTR 3 XP_016881577.1
XM_017026089.1 2405 UTR 3 XP_016881578.1
XM_017026090.1 2405 UTR 3 XP_016881579.1
XM_017026091.1 2405 UTR 3 XP_016881580.1
XM_017026092.1 2405 UTR 3 XP_016881581.1

View Full Product Details