Product Details

SNP ID
rs145814903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79904019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGGTGCACGGCGTGGGGCGCC[A/G]GCTTCTGGGGGTGGCGGGCGAAGGC
Phenotype
MIM: 605696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNG2 PubMed Links

Gene Details

Gene
KCNG2
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012283.1 815 Intron NP_036415.1
Gene
PQLC1
Gene Name
PQ loop repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146343.1 815 UTR 3 NP_001139815.1
NM_001146345.1 815 Missense Mutation CCG,CTG P240L NP_001139817.1
NM_025078.4 815 Missense Mutation CCG,CTG P258L NP_079354.2
XM_005266770.2 815 UTR 3 XP_005266827.1
XM_011526188.1 815 Missense Mutation CCG,CTG P140L XP_011524490.1
XM_017025997.1 815 Missense Mutation CCG,CTG P283L XP_016881486.1
XM_017025998.1 815 Missense Mutation CCG,CTG P265L XP_016881487.1
XM_017025999.1 815 Intron XP_016881488.1
XM_017026000.1 815 Intron XP_016881489.1
XM_017026001.1 815 Missense Mutation CCG,CTG P177L XP_016881490.1
XM_017026002.1 815 Intron XP_016881491.1
XM_017026003.1 815 UTR 3 XP_016881492.1
XM_017026004.1 815 UTR 3 XP_016881493.1
XM_017026005.1 815 Missense Mutation CCG,CTG P140L XP_016881494.1

View Full Product Details