Product Details

SNP ID
rs146944735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14752974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATTTGTTAATGGTGGCAACA[C/T]TGCTGTCCTATGGTGCAGTCATCGA
Phenotype
MIM: 616565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD30B PubMed Links

Gene Details

Gene
ANKRD30B
Gene Name
ankyrin repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145029.1 557 Silent Mutation CTG,TTG L158L NP_001138501.1
XM_006722325.3 557 Intron XP_006722388.1
XM_011525662.2 557 Silent Mutation CTG,TTG L158L XP_011523964.1
XM_011525664.2 557 Intron XP_011523966.1
XM_011525665.2 557 Silent Mutation CTG,TTG L158L XP_011523967.1
XM_011525666.2 557 Silent Mutation CTG,TTG L158L XP_011523968.1

View Full Product Details