Product Details

SNP ID
rs147713692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:69409679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTACTTGGAAAAATAGATGCA[A/G]TATTTGTCTAAAATTGGAAAGTTAG
Phenotype
MIM: 611402
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOK6 PubMed Links

Gene Details

Gene
DOK6
Gene Name
docking protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152721.5 Intron NP_689934.2
XM_011525875.2 Intron XP_011524177.1
XM_017025610.1 Intron XP_016881099.1
XM_017025611.1 Intron XP_016881100.1

View Full Product Details