Product Details

SNP ID
rs147910406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76979012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGGTTTGTGTCTGGAGTTTTG[G/T]TCTTTGTAACTCTCTCATCATCGAG
Phenotype
MIM: 159430 MIM: 604760
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MBP PubMed Links

Gene Details

Gene
MBP
Gene Name
myelin basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025081.1 2067 UTR 3 NP_001020252.1
NM_001025090.1 2067 UTR 3 NP_001020261.1
NM_001025092.1 2067 UTR 3 NP_001020263.1
NM_001025100.1 2067 Intron NP_001020271.1
NM_001025101.1 2067 UTR 3 NP_001020272.1
NM_002385.2 2067 UTR 3 NP_002376.1
XM_011526009.2 2067 Intron XP_011524311.1
XM_017025778.1 2067 Intron XP_016881267.1
XM_017025779.1 2067 Intron XP_016881268.1
XM_017025780.1 2067 Intron XP_016881269.1
Gene
ZNF236
Gene Name
zinc finger protein 236
There are no transcripts associated with this gene.

View Full Product Details