Product Details

SNP ID
rs149358681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:65752954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTTAATCACTTTACAGACTCA[C/T]AAGTCTTCTTAAGTCTACTTTTATT
Phenotype
MIM: 605806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH7 PubMed Links

Gene Details

Gene
CDH7
Gene Name
cadherin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317214.1 Intron NP_001304143.1
NM_004361.3 Intron NP_004352.2
NM_033646.2 Intron NP_387450.1
XM_017025523.1 Intron XP_016881012.1

View Full Product Details