Product Details

SNP ID
rs137926229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52065532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCTCTCCAGTATGAATACTCC[A/G]ATGACGTGCGAGGCCTGAGCGATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF841 PubMed Links

Gene Details

Gene
ZNF841
Gene Name
zinc finger protein 841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136499.1 4405 Missense Mutation CGG,TGG R784W NP_001129971.1
NM_001321349.1 4405 Missense Mutation CGG,TGG R668W NP_001308278.1
XM_011526833.2 4405 Intron XP_011525135.1
XM_017026652.1 4405 Intron XP_016882141.1
XM_017026653.1 4405 Missense Mutation CGG,TGG R785W XP_016882142.1
XM_017026654.1 4405 Missense Mutation CGG,TGG R785W XP_016882143.1
XM_017026655.1 4405 Intron XP_016882144.1
XM_017026656.1 4405 Missense Mutation CGG,TGG R784W XP_016882145.1
XM_017026657.1 4405 Intron XP_016882146.1
XM_017026658.1 4405 Intron XP_016882147.1
XM_017026659.1 4405 Missense Mutation CGG,TGG R786W XP_016882148.1
XM_017026660.1 4405 Intron XP_016882149.1
XM_017026661.1 4405 Intron XP_016882150.1
XM_017026662.1 4405 Missense Mutation CGG,TGG R784W XP_016882151.1
XM_017026663.1 4405 Intron XP_016882152.1

View Full Product Details