Product Details
- SNP ID
-
hCV165246677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8697621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCACGTGGGTCTCGGCTGGCAGAG[C/T]GCGCAGCAGCTCTGCCCGGAAGCGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTL9
PubMed Links
Gene Details
- Gene
- ACTL9
- Gene Name
- actin like 9
There are no transcripts associated with this gene.
View Full Product Details