Product Details

SNP ID
rs138148689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39605261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCCGTTTCCGAGTGCACTTTG[G/T]CAATCATGTGGTCATGAACAGGCGT
Phenotype
MIM: 608717
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LGALS13 PubMed Links

Gene Details

Gene
LGALS13
Gene Name
galectin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013268.2 216 Missense Mutation GGC,GTC G59V NP_037400.1
XM_011526874.2 216 Missense Mutation GGC,GTC G52V XP_011525176.1
XM_017026715.1 216 Missense Mutation GGC,GTC G90V XP_016882204.1

View Full Product Details