Product Details

SNP ID
rs138436841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57456013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGGGGATCTAATCTTGATCTC[A/C]ATCCACCTGCATATACTGGGGTTGA
Phenotype
MIM: 605234
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VN1R1 PubMed Links

Gene Details

Gene
VN1R1
Gene Name
vomeronasal 1 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020633.3 727 Missense Mutation ATG,ATT M158I NP_065684.1
Gene
ZNF749
Gene Name
zinc finger protein 749
There are no transcripts associated with this gene.

Gene
ZNF772
Gene Name
zinc finger protein 772
There are no transcripts associated with this gene.

View Full Product Details