Product Details

SNP ID
rs138505126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48446421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCTTTGACATAGTCCGGGATC[A/G]CCTGGGAGACAACCGGACAGAGCTT
Phenotype
MIM: 602717 MIM: 610597 MIM: 603953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN2D PubMed Links

Gene Details

Gene
GRIN2D
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
There are no transcripts associated with this gene.

Gene
GRWD1
Gene Name
glutamate rich WD repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031485.3 457 Missense Mutation CAC,CGC H75R NP_113673.3
Gene
KCNJ14
Gene Name
potassium voltage-gated channel subfamily J member 14
There are no transcripts associated with this gene.

View Full Product Details