Product Details

SNP ID
rs138632069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10317486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCTTCCGCTTCAGGTAGGAGT[C/G]CGCGTAGTGGCCGCCGAGGCCCTGG
Phenotype
MIM: 614585 MIM: 609950 MIM: 611639
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FDX1L PubMed Links

Gene Details

Gene
FDX1L
Gene Name
ferredoxin 1 like
There are no transcripts associated with this gene.

Gene
RAVER1
Gene Name
ribonucleoprotein, PTB binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133452.2 2319 Missense Mutation CAC,GAC H747D NP_597709.2
XM_011527671.1 2319 Missense Mutation CAC,GAC H722D XP_011525973.1
Gene
ZGLP1
Gene Name
zinc finger, GATA-like protein 1
There are no transcripts associated with this gene.

View Full Product Details