Product Details
- SNP ID
-
rs138644874
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51125367 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGAAGTATAAAATGGAATTATAA[A/C]CATCACCGGCTCTCTGTGAATGTGA
- Phenotype
-
MIM: 605640
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SIGLEC9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16988910] are located under a probe and SNP(s) [rs2075803] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SIGLEC9
- Gene Name
- sialic acid binding Ig like lectin 9
View Full Product Details