Product Details

SNP ID
rs138758439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54103138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCGGCGCCTTCCTCAAGAATG[C/G]CTGGGACAAGGAGCCAGTGCTGGTC
Phenotype
MIM: 603832 MIM: 606862 MIM: 609519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004542.3 479 Missense Mutation GCC,GGC A12G NP_004533.1
XM_017026833.1 479 Missense Mutation GCC,GGC A12G XP_016882322.1
Gene
OSCAR
Gene Name
osteoclast associated, immunoglobulin-like receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282349.1 479 Intron NP_001269278.1
NM_001282350.1 479 Intron NP_001269279.1
NM_130771.4 479 Intron NP_570127.3
NM_133168.4 479 Intron NP_573398.2
NM_133169.4 479 Intron NP_573399.2
NM_206818.2 479 Intron NP_996554.2
Gene
TFPT
Gene Name
TCF3 fusion partner
There are no transcripts associated with this gene.

View Full Product Details