Product Details
- SNP ID
-
rs139260207
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37669375 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAAGCTCTACCACATTCTTTAC[A/T]TTCATAGGGTTTTTCACCAGTGTGA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF781
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73932934] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF781
- Gene Name
- zinc finger protein 781
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152605.3 |
1516 |
Missense Mutation |
AAA,AAT |
K258N |
NP_689818.2 |
View Full Product Details