Product Details

SNP ID
rs139657571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46775514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAAGGTCCCTCCCGGGGTTTAC[A/G]TGACTGATTCCTTCTCAGAGGCGAC
Phenotype
MIM: 109190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC1A5 PubMed Links

Gene Details

Gene
SLC1A5
Gene Name
solute carrier family 1 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145144.1 1296 Missense Mutation ACG,ATG T313M NP_001138616.1
NM_001145145.1 1296 Missense Mutation ACG,ATG T339M NP_001138617.1
NM_005628.2 1296 Missense Mutation ACG,ATG T541M NP_005619.1
XM_005259167.4 1296 Intron XP_005259224.1

View Full Product Details