Product Details

SNP ID
rs139693115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56845827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTACTAATATTATTAATCTCTC[C/T]ATGTTTCTCCATCAGTTTTGCTAAA
Phenotype
MIM: 601483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIMT1 PubMed Links

Gene Details

Gene
MIMT1
Gene Name
MER1 repeat containing imprinted transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
PEG3
Gene Name
paternally expressed 3
There are no transcripts associated with this gene.

Gene
ZIM2
Gene Name
zinc finger imprinted 2
There are no transcripts associated with this gene.

View Full Product Details