Product Details

SNP ID
rs139802061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1581174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACGCCGGGGTTCTTCTCCACGG[C/T]GGCCGAGAGCTGTCCCGTGATGGGC
Phenotype
MIM: 603573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD3 PubMed Links

Gene Details

Gene
MBD3
Gene Name
methyl-CpG binding domain protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281453.1 725 Missense Mutation ACC,GCC T199A NP_001268382.1
NM_001281454.1 725 Missense Mutation ACC,GCC T167A NP_001268383.1

View Full Product Details