Product Details

SNP ID
rs140078852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43584588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAAGTCTGTATTAGTTGCTTTT[A/G]CTACATAACAAACCACTCCAAAATT
Phenotype
MIM: 194360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007561.2 9466 UTR 3 NP_001007562.1
XM_005258515.3 9466 Intron XP_005258572.1
Gene
PINLYP
Gene Name
phospholipase A2 inhibitor and LY6/PLAUR domain containing
There are no transcripts associated with this gene.

Gene
XRCC1
Gene Name
X-ray repair cross complementing 1
There are no transcripts associated with this gene.

View Full Product Details